MilliporeSigma
  • Use of uric acid-lowering agents limits experimental cyclosporine nephropathy.
Use of uric acid-lowering agents limits experimental cyclosporine nephropathy.
Nephron. Experimental nephrology (2011-12-01)
Fernanda Cristina Mazali, Richard J Johnson, Marilda Mazzali
ABSTRACT
Hyperuricemia frequently complicates cyclosporine (CsA) therapy. Previous studies have shown that hyperuricemia exacerbates interstitial and vascular lesions in the cyclosporine model. We tested the hypothesis that normalization of uric acid could prevent the development of cyclosporine toxicity. CsA nephropathy was induced by administering CsA (15 mg/kg/day) for 7 weeks to rats on a low salt diet (CsA group). The effect of preventing hyperuricemia was determined by concomitant treatment with a xanthine oxidase inhibitor, allopurinol (CsAALP), or with a uricosuric, benzbromarone (CsABENZ), in drinking water. Control groups included vehicle-treated rats. CsA-treated rats developed mild hyperuricemia with arteriolar hyalinosis, tubular atrophy, striped interstitial fibrosis, increased cell proliferation and decreased VEGF expression. Treatment with allopurinol or benzbromarone limited renal disease, with reduced interstitial fibrosis, cell proliferation, macrophage infiltration, osteopontin expression and arteriolar hyalinosis, in association with restoration of VEGF expression. Both drugs provided comparable protection. An increase in uric acid exacerbates CsA nephropathy in the rat. Concomitant treatment with allopurinol or benzbromarone reduced the severity of renal disease. The similar protection observed with both drugs suggests that the effect is associated more with lowering uric acid levels than the antioxidant effect of allopurinol.
Product Description
Benzbromarone, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzbromarone, analytical standard