MilliporeSigma
Chemical biology of homocysteine thiolactone and related metabolites.
Advances in clinical chemistry (2011-12-01)
Hieronim Jakubowski, Rafał Głowacki
ABSTRACT
Protein-related homocysteine (Hcy) metabolism produces Hcy-thiolactone, N-Hcy-protein, and N epsilon-homocysteinyl-lysine (N epsilon-Hcy-Lys). Hcy-thiolactone is generated in an error-editing reaction in protein biosynthesis when Hcy is erroneously selected in place of methionine by methionyl-tRNA synthetase. Hcy-thiolactone, an intramolecular thioester, is chemically reactive and forms isopeptide bonds with protein lysine residues in a process called N-homocysteinylation, which impairs or alters the protein's biological function. The resulting protein damage is exacerbated by a thiyl radical-mediated oxidation. N-Hcy-proteins undergo structural changes leading to aggregation and amyloid formation. These structural changes generate proteins, which are toxic and which induce an autoimmune response. Proteolytic degradation of N-Hcy-proteins generates N epsilon-Hcy-Lys. Levels of Hcy-thiolactone, N-Hcy-protein, and N epsilon-Hcy-Lys increase under pathological conditions in humans and mice and have been linked to cardiovascular and brain disorders. This chapter reviews fundamental biological chemistry of Hcy-thiolactone, N-Hcy-protein, and N epsilon-Hcy-Lys and discusses their clinical significance.
Sigma-Aldrich
DL-Homocysteine thiolactone hydrochloride, ≥99.0% (AT)