MilliporeSigma
  • A simple "molecular beacon"-based fluorescent sensing strategy for sensitive and selective detection of mercury (II).
A simple "molecular beacon"-based fluorescent sensing strategy for sensitive and selective detection of mercury (II).
Chemical communications (Cambridge, England) (2011-10-12)
Huifeng Xu, Xi Zhu, Hongzhi Ye, Lishuang Yu, Xianxiang Liu, Guonan Chen
ABSTRACT
A novel fluorescent sensor for the detection of Hg(2+) in aqueous media was developed. The method takes advantages of the highly selective thymine-Hg(2+)-thymine coordination and the sensitive "signal-on" structure-switching molecular beacon.
Product Description
Supelco
Butter Yellow, analytical standard