MilliporeSigma
  • Shewanelia putrefaciens: a rare microbial agent associated with a non-healing ulcer in a leprosy patient.
Shewanelia putrefaciens: a rare microbial agent associated with a non-healing ulcer in a leprosy patient.
Indian journal of leprosy (2011-03-26)
D S Bhalerao, A G Kinikar, S B Roushani, V X Franklin
ABSTRACT
Female aged 55 years presented with signs and symptoms of borderline lepromatous leprosy and presence of a non-healing ulcer and multiple haemorrhagic blisters over dorsum of both feet. Discharge from the various lesions was subjected to microbiological examination and an unusual organism Shewanella purtefaciens was isolated which was sensitive to most routine antibiotics. Patient responded well to cephadroxil therapy with uneventful and complete healing of ulcer and blisters.
Product Description
Supelco
Cefadroxil, analytical standard