MilliporeSigma
  • Monitoring of antisolvent crystallization of sodium scutellarein by combined FBRM-PVM-NIR.
Monitoring of antisolvent crystallization of sodium scutellarein by combined FBRM-PVM-NIR.
Journal of pharmaceutical sciences (2011-01-20)
Xuesong Liu, Di Sun, Feng Wang, Yongjiang Wu, Yong Chen, Longhu Wang
ABSTRACT
Antisolvent crystallization can be used as an alternative to cooling or evaporation for the separation and purification of solid product in the pharmaceutical industry. To improve the process understanding of antisolvent crystallization, the use of in-line tools is vital. In this study, the process analytical technology (PAT) tools including focused beam reflectance measurement (FBRM), particle video microscope (PVM), and near-infrared spectroscopy (NIRS) were utilized to monitor antisolvent crystallization of sodium scutellarein. FBRM was used to monitor chord count and chord length distribution of sodium scutellarein particles in the crystallizer, and PVM, as an in-line video camera, provided pictures imaging particle shape and dimension. In addition, a quantitative model of PLS was established by in-line NIRS to detect the concentration of sodium scutellarein in the solvent and good calibration statistics were obtained (r(2) = 0.976) with the residual predictive deviation value of 11.3. The discussion over sensitivities, strengths, and weaknesses of the PAT tools may be helpful in selection of suitable PAT techniques. These in-line techniques eliminate the need for sample preparation and offer a time-saving approach to understand and monitor antisolvent crystallization process.
Sigma-Aldrich
Scutellarein, ≥98% (HPLC)