MilliporeSigma
  • Synthesis and antituberculosis activity of some new pyridazine derivatives. Part II.
Synthesis and antituberculosis activity of some new pyridazine derivatives. Part II.
European journal of medicinal chemistry (2010-09-04)
Dorina Mantu, Mihaela Cătălina Luca, Costel Moldoveanu, Gheorghita Zbancioc, Ionel I Mangalagiu
ABSTRACT
A series of eighteen novel compounds with pyridazine moiety were synthesized and their in vitro antituberculosis activities have been evaluated. A fast, general, and facile method for preparation of pyridazine derivatives in moderate to excellent yields is presented. Three compounds were found to be moderate active against Mycobacterium tuberculosis. Correlation of structure-biological activity has been done.
Sigma-Aldrich
Pyridazine, 98%