MilliporeSigma
  • Manganese catalyzed cis-dihydroxylation of electron deficient alkenes with H(2)O(2).
Manganese catalyzed cis-dihydroxylation of electron deficient alkenes with H(2)O(2).
Organic & biomolecular chemistry (2010-08-18)
Pattama Saisaha, Dirk Pijper, Ruben P van Summeren, Rob Hoen, Christian Smit, Johannes W de Boer, Ronald Hage, Paul L Alsters, Ben L Feringa, Wesley R Browne
ABSTRACT
A practical method for the multigram scale selective cis-dihydroxylation of electron deficient alkenes such as diethyl fumarate and N-alkyl and N-aryl-maleimides using H(2)O(2) is described. High turnovers (>1000) can be achieved with this efficient manganese based catalyst system, prepared in situ from a manganese salt, pyridine-2-carboxylic acid, a ketone and a base, under ambient conditions. Under optimized conditions, for diethyl fumarate at least 1000 turnovers could be achieved with only 1.5 equiv. of H(2)O(2) with d/l-diethyl tartrate (cis-diol product) as the sole product. For electron rich alkenes, such as cis-cyclooctene, this catalyst provides for efficient epoxidation.
Sigma-Aldrich
Diethyl fumarate, 98%