MilliporeSigma
  • Grafting of poly(ethylene glycol) onto poly(acrylic acid)-coated glass for a protein-resistant surface.
Grafting of poly(ethylene glycol) onto poly(acrylic acid)-coated glass for a protein-resistant surface.
Analytical chemistry (2006-04-18)
Tetsuichi Wazawa, Yoshiko Ishizuka-Katsura, So Nishikawa, Atsuko Hikikoshi Iwane, Shigeru Aoyama
ABSTRACT
The surface of solid glass supports for samples in optical microscopy and for biosensors needs to be protein-resistant. A coating of a poly(ethylene glycol) monomethyl ether (mPEG) on the surface of the glass is one promising method for preventing the nonspecific adsorption of proteins. In this study, we have developed a novel technique for achieving an optimal coverage of a glass surface with mPEG to prevent protein adhesion. A clean glass substrate previously treated with (3-aminopropyl)dimethylethoxysilane (APDMES) was treated sequentially with poly(acrylic acid) and subsequently a primary amine derivative of mPEG in the presence of 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide. The resultant glass surface was demonstrated to be highly protein-resistant, and the adsorption of bovine serum albumin decreased to only a few percentage points of that on a glass surface treated with APDMES alone. Furthermore, to extend the present method, we also prepared a glass substrate on which biotinylated poly(ethylene glycol) was cografted with mPEG, and biotinylated myosin subfragment-1 (biotin-S1) was subsequently immobilized on this substrate by biotin/avidin chemistry. Actin filaments were observed to glide on the biotin-S1-coated glass surface in the presence of ATP, and thus, the method is capable of immobilizing the protein specifically without any loss in its biological function.
Sigma-Aldrich
3-Dimethylamino-1-propanol, 99%