MilliporeSigma
  • Facile formation of an intrastrand cross-link lesion between cytosine and guanine upon pyrex-filtered UV light irradiation of d((Br)CG) and duplex DNA containing 5-bromocytosine.
Facile formation of an intrastrand cross-link lesion between cytosine and guanine upon pyrex-filtered UV light irradiation of d((Br)CG) and duplex DNA containing 5-bromocytosine.
Journal of the American Chemical Society (2004-05-27)
Yu Zeng, Yinsheng Wang
ABSTRACT
Pyrex-filtered UV light irradiation of d(BrCG) and 5-bromocytosine-containing duplex DNA leads to facile formation of a cross-link lesion between the C5 carbon atom of cytosine and the C8 carbon atom of its adjacent guanine. A similar cross-link lesion has been previously found in the X-ray irradiation mixture of d(CGTA).