MilliporeSigma
Complex formation of isocytosine tautomers with PdII and PtII.
Inorganic chemistry (2004-05-25)
Deepali Gupta, Markus Huelsekopf, Marta Morell Cerdà, Ralf Ludwig, Bernhard Lippert
ABSTRACT
Isocytosine (ICH) exists in solution as two major tautomers, the keto form with N1 carrying a proton (1a) and the keto form with N3 being protonated (1b). In water, 1a and 1b exist in equilibrium with almost equal amounts of both forms present. Reactions with a series of Pd(II) and Pt(II) am(m)ine species such as (dien)Pd(II), (dien)Pt(II), and trans-(NH(3))(2)Pt(II) reveal, however, a distinct preference of these metals for the N3 site, as determined by (1)H NMR spectroscopy. Individual species have been identified by the pD dependence of the ICH resonances. pK(a) values (calculated for H(2)O) for deprotonation of the individual tautomers complexes are 6.5 and 6.4 for the N3 linkage isomers of dienPd(II) and dienPt(II), respectively, as well as 6.2 and 6.0 for the N1 linkage isomers. The dimetalated species [(dienM)(2)(IC-N1,N3)](3+) (M = Pd(II) or Pt(II)) are insensitive over a wide range of pD. The crystal structure analysis of [(dien)Pd(ICH-N3)](NO(3))(2) is reported. Ab initio calculations have been performed for tautomer compounds of composition [(NH(3))(3)Pt(ICH)](2+), cis- and trans-[(NH(3))(2)PtCl(ICH)](+), as well as trans-[(NH(3))(2)Pt(ICH)(2)](2+). Without exception, N3 linkage isomers are more stable, in agreement with experimental findings. As to the reasons for this binding preference, an NBO (natural bond orbital) analysis for [(NH(3))(3)Pt(ICH-N3)](2+)strongly suggests that intramolecular hydrogen bonding between trans-positioned NH(3) ligands and the two exocyclic groups of the ICH is of prime importance. The calculations furthermore show a marked pyramidalization of the NH(2) group of ICH in the complex once the heterocyclic ligand forms a dihedral angle <90 degrees with the Pt coordination plane.