MilliporeSigma
  • Gene cluster of Arthrobacter ilicis Ru61a involved in the degradation of quinaldine to anthranilate: characterization and functional expression of the quinaldine 4-oxidase qoxLMS genes.
Gene cluster of Arthrobacter ilicis Ru61a involved in the degradation of quinaldine to anthranilate: characterization and functional expression of the quinaldine 4-oxidase qoxLMS genes.
The Journal of biological chemistry (2003-05-06)
Katja Parschat, Bernhard Hauer, Reinhard Kappl, Roswitha Kraft, Jurgen Huttermann, Susanne Fetzner
ABSTRACT
A genetic analysis of the anthranilate pathway of quinaldine degradation was performed. A 23-kb region of DNA from Arthrobacter ilicis Rü61a was cloned into the cosmid pVK100. Although Escherichia coli clones containing the recombinant cosmid did not transform quinaldine, cosmids harboring the 23-kb region, or a 10.8-kb stretch of this region, conferred to Pseudomonas putida KT2440 the ability to cometabolically convert quinaldine to anthranilate. The 10.8-kb fragment thus contains the genes coding for quinaldine 4-oxidase (Qox), 1H-4-oxoquinaldine 3-monooxygenase, 1H-3-hydroxy-4-oxoquinaldine 2,4-dioxygenase, and N-acetylanthranilate amidase. The qoxLMS genes coding for the molybdopterin cytosine dinucleotide-(MCD-), FeSI-, FeSII-, and FAD-containing Qox were inserted into the expression vector pJB653, generating pKP1. Qox is the first MCD-containing enzyme to be synthesized in a catalytically fully competent form by a heterologous host, P. putida KT2440 pKP1; the catalytic properties and the UV-visible and EPR spectra of Qox purified from P. putida KT2440 pKP1 were essentially like those of wild-type Qox. This provides a starting point for the construction of protein variants of Qox by site-directed mutagenesis. Downstream of the qoxLMS genes, a putative gene whose deduced amino acid sequence showed 37% similarity to the cofactor-inserting chaperone XdhC was located. Additional open reading frames identified on the 23-kb segment may encode further enzymes (a glutamyl tRNA synthetase, an esterase, two short-chain dehydrogenases/reductases, an ATPase belonging to the AAA family, a 2-hydroxyhepta-2,4-diene-1,7-dioate isomerase/5-oxopent-3-ene-1,2,5-tricarboxylate decarboxylase-like protein, and an enzyme of the mandelate racemase group) and hypothetical proteins involved in transcriptional regulation, and metabolite transport.
Sigma-Aldrich
Quinaldine, ≥95.0% (GC)
Supelco
Quinaldine, analytical standard, ≥90% (GC)