MilliporeSigma
  • Reactions of morphine derivatives with phenyliodo(III)diacetate (PIDA): synthesis of new morphine analogues.
Reactions of morphine derivatives with phenyliodo(III)diacetate (PIDA): synthesis of new morphine analogues.
Current medicinal chemistry (2001-04-03)
S Garadnay, P Herczegh, S Makleit, T Friedmann, P Riba, S Fürst
ABSTRACT
The reactions of morphine and its derivatives with phenyliodo(III)diacetate (PIDA) have been studied. This methodology has not been introduced to morphine alkaloids, despite the fact that such a strategy would ensure dearomatization of the electrophilic aromatic ring of morphine derivatives leading to nucleophilic ortho-quinoidal structures with potential pharmacological interest. The products, formed in regio- and diastereoselective or diastereospecific reactions, carry mixed-acetal or 1,3-dioxolane moieties. At low concentrations 6a has mu-opioid agonist character but in higher concentrations showed a non receptorial antagonist effect on isolated mouse vas deferens.
Sigma-Aldrich
(Diacetoxyiodo)benzene, 98%