MilliporeSigma
  • Cloning and sequencing of inulinase and β-fructofuranosidase genes of a deep-sea Microbulbifer species and properties of recombinant enzymes.
Cloning and sequencing of inulinase and β-fructofuranosidase genes of a deep-sea Microbulbifer species and properties of recombinant enzymes.
Applied and environmental microbiology (2012-01-31)
Tohru Kobayashi, Kohsuke Uchimura, Shigeru Deguchi, Koki Horikoshi
ABSTRACT
An inulinase-producing Microbulbifer sp. strain, JAM-3301, was isolated from a deep-sea sediment. An inulin operon that contained three open reading frames was cloned and sequenced. Two of the three genes were expressed. One product was an endo-inulinase, and the other was a β-fructofuranosidase. Both enzymes worked together to effectively degrade inulin.
Sigma-Aldrich
Inulinase from Aspergillus niger, lyophilized, powder, brown-gray, ~25 U/mg