MilliporeSigma
  • A new method of synthesis of alkyl beta-glycosides using sucrose as sugar donor.
A new method of synthesis of alkyl beta-glycosides using sucrose as sugar donor.
Bioscience, biotechnology, and biochemistry (2008-09-09)
Kuniki Kino, Ryoko Satake, Takayuki Morimatsu, Shoko Kuratsu, Yu Shimizu, Masaru Sato, Kohtaro Kirimura
ABSTRACT
Cellobiose phosphorylase from Clostridium thermocellum catalyzed the beta-anomer-selective synthesis of alkyl glucosides from cellobiose. Synthesis of alkyl beta-glucoside from inexpensive sucrose using cellobiose phosphorylase and sucrose phosphorylase from Pseudomonas saccharophilia was investigated. By combined use of these two phosphorylases, alkyl beta-glucoside was anomer-selectively synthesized from sucrose and alkyl alcohol.
Sigma-Aldrich
Sucrose Phosphorylase, recombinant, expressed in E. coli, lyophilized powder, ≥45 units/mg solid