MilliporeSigma
  • [Pharmacological correction of energy metabolism disturbance in the inflammatory-dystrophic process in the periodontium].
[Pharmacological correction of energy metabolism disturbance in the inflammatory-dystrophic process in the periodontium].
Eksperimental'naia i klinicheskaia farmakologiia (2006-09-26)
V D Luk''ianchuk, O A Shpulina
ABSTRACT
The influence of lipoic acid on the energy homeostasis in rats with chronic generalized parodontitis has been investigated. Lipoic acid possesses the energy-saving activity, which is related to an increase in oxidative phosphorylation efficiency.
Sigma-Aldrich
Creatininase from microorganisms, lyophilized powder, 100-300 units/mg protein