MilliporeSigma
  • Hypomorphic pathogenic variant in SFTPB leads to adult pulmonary fibrosis.
Hypomorphic pathogenic variant in SFTPB leads to adult pulmonary fibrosis.
European journal of human genetics : EJHG (2023-06-29)
Tifenn Desroziers, Grégoire Prévot, Aurore Coulomb, Valérie Nau, Florence Dastot-Le Moal, Philippe Duquesnoy, Mélanie Héry, Aurélie Le Borgne, Serge Amselem, Marie Legendre, Nadia Nathan
ABSTRACT
Biallelic pathogenic variants in the surfactant protein (SP)-B gene (SFTPB) have been associated with fatal forms of interstitial lung diseases (ILD) in newborns and exceptional survival in young children. We herein report the cases of two related adults with pulmonary fibrosis due to a new homozygous SFTPB pathogenic variant, c.582G>A p.(Gln194=). In vitro transcript studies showed that this SFTPB synonymous pathogenic variant induces aberrant splicing leading to three abnormal transcripts with the preservation of the expression of a small proportion of normal SFTPB transcripts. Immunostainings on lung biopsies of the proband showed an almost complete loss of SP-B expression. This hypomorphic splice variant has thus probably allowed the patients' survival to adulthood while inducing an epithelial cell dysfunction leading to ILD. Altogether, this report shows that SFTPB pathogenic variants should be considered in atypical presentations and/or early-onset forms of ILD particularly when a family history is identified.
Sigma-Aldrich
Anti-SFTPB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution