MilliporeSigma
  • Fate-mapping analysis of cochlear cells expressing Atoh1 mRNA via a new Atoh13*HA-P2A-Cre knockin mouse strain.
Fate-mapping analysis of cochlear cells expressing Atoh1 mRNA via a new Atoh13*HA-P2A-Cre knockin mouse strain.
Developmental dynamics : an official publication of the American Association of Anatomists (2022-01-18)
Shuting Li, Ting Fan, Chao Li, Yunfeng Wang, Jian Li, Zhiyong Liu
ABSTRACT
Atoh1 is recognized to be essential for cochlear hair cell (HC) development. However, Atoh1 temporal and spatial expression patterns remain widely debated. Here, we aimed to obtain evidence to resolve the controversies regarding Atoh1 expression by generating a new knockin mouse strain: Atoh13*HA-P2A-Cre . Fate-mapping analysis of Atoh13*HA-P2A-Cre/+ ; Rosa26-CAG-LSL-tdTomato (Ai9)/+ mice enabled us to concurrently characterize the temporal expression of Atoh1 protein (through HA-tag immunostaining) and visualize the cells expressing Atoh1 mRNA (as tdTomato+ cells). Our findings show that whereas Atoh1 mRNA expression is rapidly turned on in early cochlear progenitors, Atoh1 protein is only detected in differentiating HCs or progenitors just committed to the HC fate. Cre activity is also stronger in Atoh13*HA-P2A-Cre/+ than in previous mouse models, because almost all cochlear HCs and nearby supporting cells here are tdTomato+. Furthermore, tdTomato, but not HA, is expressed in middle and apical spiral ganglion neurons. Collectively, our findings indicate that Atoh13*HA-P2A-Cre can serve as a powerful genetic model in the developmental biology field.
Sigma-Aldrich
Anti-MAFB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution