MilliporeSigma
  • Palladium(II) chloride catalyzed selective acetylation of alcohols with vinyl acetate.
Palladium(II) chloride catalyzed selective acetylation of alcohols with vinyl acetate.
Chemical communications (Cambridge, England) (2004-04-30)
J W J Bosco, Anil K Saikia
ABSTRACT
PdCl(2) can catalyze the acetylation of primary and secondary alcohols with vinyl acetate. The reaction is selective and mild with high yields. Tertiary alcohols, phenols and amines are unaffected under these reaction conditions.
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%