MilliporeSigma
  • The novel P2X7 receptor antagonist PKT100 improves cardiac function and survival in pulmonary hypertension by direct targeting of the right ventricle.
The novel P2X7 receptor antagonist PKT100 improves cardiac function and survival in pulmonary hypertension by direct targeting of the right ventricle.
American journal of physiology. Heart and circulatory physiology (2020-05-30)
Thomas Hansen, Keyvan Karimi Galougahi, Marie Besnier, Elijah Genetzakis, Michael Tsang, Meghan Finemore, James O'Brien-Brown, Belinda A Di Bartolo, Michael Kassiou, Kristen J Bubb, Gemma A Figtree
ABSTRACT
In pulmonary hypertension (PH) a proinflammatory milieu drives pulmonary vascular remodeling, maladaptive right ventricular (RV) remodeling, and right-sided heart failure. There is an unmet need for RV-targeted pharmaco-therapies to improve mortality. Targeting of the P2X7 receptor (P2X7R) reduces pulmonary pressures; however, its effects on the RV are presently unknown. We investigated the effect of P2X7 receptor (P2X7R) inhibition on the pulmonary vasculature and RV remodeling using the novel P2X7R antagonist PKT100. C57BL/6 mice were administered intratracheal bleomycin or saline and treated with PKT100 (0.2 mg·kg-1·day-1) or DMSO vehicle. RV was assessed by right heart catheterization and echocardiography, 21 days posttreatment. Cytokines in serum and bronchoalveolar lavage fluid (BALF) were analyzed by ELISA and flow cytometry. Lungs and hearts were analyzed histologically for pulmonary vascular and RV remodeling. Focused-PCR using genes involved in RV remodeling was performed. Right ventricular systolic pressure (RVSP) was elevated in bleomycin-treated mice (30.2 ± 1.1; n = 7) compared with control mice (23.5 ± 1.0; n = 10; P = 0.008). PKT100 treatment did not alter RVSP (32.4 ± 1.8; n = 9), but it substantially improved survival (93% vs. 57% DMSO). There were no differences between DMSO and PKT100 bleomycin mice in pulmonary inflammation or remodeling. However, RV hypertrophy was reduced in PKT100 mice. Bleomycin decreased echocardiographic surrogates of RV systolic performance, which were significantly improved with PKT100. Four genes involved in RV remodeling (RPSA, Rplp0, Add2, and Scn7a) were differentially expressed between DMSO and PKT100-treated groups. The novel P2X7R inhibitor, PKT100, attenuates RV hypertrophy and improves RV contractile function and survival in a mouse model of PH independently of effects on the pulmonary vasculature. PKT100 may improve ventricular response to increased afterload and merits further investigation into the potential role of P2X7R antagonists as direct RV-focused therapies in PH.NEW & NOTEWORTHY This study demonstrates the therapeutic potential for right-sided heart failure of a novel inhibitor of the P2X7 receptor (P2X7R). Inflammatory signaling and right ventricular function were improved in a mouse model of pulmonary fibrosis with secondary pulmonary hypertension when treated with this inhibitor. Importantly, survival was also improved, suggesting that this inhibitor, and other P2X7R antagonists, could be uniquely effective in right ventricle (RV)-targeted therapy in pulmonary hypertension. This addresses a major limitation of current treatment options, where the significant improvements in pulmonary pressures ultimately do not prevent mortality due to RV failure.
Sigma-Aldrich
Anti-P2X7 Receptor Antibody, extracellular domain, Chemicon®, from rabbit