MilliporeSigma
  • Improved Wound Healing by Naringin Associated with MMP and the VEGF Pathway.
Improved Wound Healing by Naringin Associated with MMP and the VEGF Pathway.
Molecules (Basel, Switzerland) (2022-03-11)
Jia-Hau Yen, Wan-Ting Chio, Chia-Ju Chuang, Hsin-Ling Yang, Sheng-Teng Huang
ABSTRACT
This study aims to investigate the wound-healing effectiveness of the phenolic compound, naringin, both in vitro and in vivo. Male mice were shaved on their dorsal skin under isoflurane, a biopsy punch was made in four symmetrical circular resection windows (6 mm) to induce a wound. These excision wounds were used to study the topical effects of naringin in terms of various biochemical, molecular, and histological parameters. We observed a significant recovery in the wound area. Increased levels of MMP-2, 9, 14, TIMP-2, VEGF-A, and VEGF-R1 were induced by naringin in the HaCaT cells. The time course experiments further revealed that levels of VEGF-A and B increased within 36 h; whereas levels of VEGF-C decreased. In line with this, VEGF-R3 levels, but not VEGF-R1 and 2 levels, increased soon after stimulation; although the increase subsided after 36 h. Additionally, naringin cream upregulated wound healing in vitro. The blockage of VEGF by Bevacizumab abolished the function of naringin cream on cell migration. Histological alterations in the wounded skin were restored by naringin cream, which accelerated wound healing via upregulated expression of growth factors (VEGF-A, B, and C and VEGF-R3), and thus increased MMP-2, 9, 14 expressions.
Sigma-Aldrich
Anti-TIMP-1 Antibody, clone 7-6C1, clone 7-6C1, Chemicon®, from mouse