MilliporeSigma
  • Polystyrene Chain Growth Initiated from Dialkylzinc for Synthesis of Polyolefin-Polystyrene Block Copolymers.
Polystyrene Chain Growth Initiated from Dialkylzinc for Synthesis of Polyolefin-Polystyrene Block Copolymers.
Polymers (2020-03-07)
Tae Jin Kim, Jun Won Baek, Seung Hyun Moon, Hyun Ju Lee, Kyung Lee Park, Sung Moon Bae, Jong Chul Lee, Pyung Cheon Lee, Bun Yeoul Lee
ABSTRACT
Polyolefins (POs) are the most abundant polymers. However, synthesis of PO-based block copolymers has only rarely been achieved. We aimed to synthesize various PO-based block copolymers by coordinative chain transfer polymerization (CCTP) followed by anionic polymerization in one-pot via conversion of the CCTP product (polyolefinyl)2Zn to polyolefinyl-Li. The addition of 2 equiv t-BuLi to (1-octyl)2Zn (a model compound of (polyolefinyl)2Zn) and selective removal or decomposition of (tBu)2Zn by evacuation or heating at 130 °C afforded 1-octyl-Li. Attempts to convert (polyolefinyl)2Zn to polyolefinyl-Li were unsuccessful. However, polystyrene (PS) chains were efficiently grown from (polyolefinyl)2Zn; the addition of styrene monomers after treatment with t-BuLi and pentamethyldiethylenetriamine (PMDTA) in the presence of residual olefin monomers afforded PO-block-PSs. Organolithium species that might be generated in the pot of t-BuLi, PMDTA, and olefin monomers, i.e., [Me2NCH2CH2N(Me)CH2CH2N(Me)CH2Li, Me2NCH2CH2N(Me)Li·(PMDTA), pentylallyl-Li⋅(PMDTA)], as well as PhLi⋅(PMDTA), were screened as initiators to grow PS chains from (1-hexyl)2Zn, as well as from (polyolefinyl)2Zn. Pentylallyl-Li⋅(PMDTA) was the best initiator. The Mn values increased substantially after the styrene polymerization with some generation of homo-PSs (27-29%). The Mn values of the extracted homo-PS suggested that PS chains were grown mainly from polyolefinyl groups in [(polyolefinyl)2(pentylallyl)Zn]-[Li⋅(PMDTA)]+ formed by pentylallyl-Li⋅(PMDTA) acting onto (polyolefinyl)2Zn.
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%