MilliporeSigma
  • Treatment of malignancy-associated hypercalcemia and cachexia with humanized anti-parathyroid hormone-related protein antibody.
Treatment of malignancy-associated hypercalcemia and cachexia with humanized anti-parathyroid hormone-related protein antibody.
Seminars in oncology (2003-11-13)
Koh Sato, Etsuro Onuma, Richard C Yocum, Etsuro Ogata
ABSTRACT
Parathyroid hormone-related protein (PTHrP) plays a central role in humoral hypercalcemia of malignancy (HHM), which is one of the most frequent paraneoplastic syndromes. PTHrP produced by the tumor acts through a common PTH/PTHrP receptor to promote bone resorption, inhibit calcium excretion from the kidney, and induce hypercalcemia. Patients with HHM often develop cachexia associated with typical symptoms such as anorexia, malaise, nausea, constipation, polyuria, polydipsia, and confusion. The etiology of the cachexia is not fully understood but is thought to be caused by hypercalcemia and various cytokines such as interleukin-6, tumor necrosis factor-alpha, leukemia inhibitory factor, and others. In this study, we investigated the role of PTHrP in hypercalcemia and cachexia in HHM by using humanized anti-PTHrP antibody. A mouse monoclonal antibody that binds to PTHrP amino acid sequence 1-34 and inhibits PTHrP function has been humanized to create a specific and potent agent for the treatment of patients with HHM. The mouse monoclonal antibody has been shown to have antihypercalcemic activity against nude mice bearing human tumors. Because a mouse antibody is highly immunogenic in human patients, the complementarity-determining regions from the mouse antibody were grafted into a human antibody. The resulting humanized antibody specifically recognizes PTHrP(1-34) and neutralizes PTHrP functions in vitro and in vivo. The humanized anti-PTHrP antibody was administered intravenously to HHM model animals bearing tumors such as LC-6 human lung carcinoma. These animals showed symptoms similar to those of patients with HHM (eg, hypercalcemia and cachexia). The humanized anti-PTHrP antibody-treated animals responded with normalization of blood ionized calcium level through an improvement of bone metabolism and calcium excretion. Moreover, the treated animals also showed an improvement in body weight, ultromotivity, metabolic alkalosis, food consumption, water intake, serum phosphorus, and renal function. Consequently, the humanized antibody-treated animals experienced complete resolution of hypercalcemia and cachexia. These results suggest that the humanized antibody would be an effective and beneficial agent for patients with HHM, and that PTHrP is a major pathogenetic factor of hypercalcemia and cachexia in patients with HHM.
Sigma-Aldrich
Hypercalcemia of malignancy factor 1-40 human, ≥95% (HPLC)
Sigma-Aldrich
Hypercalcemia of malignancy factor fragment 1-34 amide human, ≥97% (HPLC)