MilliporeSigma
  • The immunophenotype of epithelial to mesenchymal transition inducing transcription factors in salivary gland adenoid cystic carcinomas.
The immunophenotype of epithelial to mesenchymal transition inducing transcription factors in salivary gland adenoid cystic carcinomas.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie (2021-04-06)
Iulia Cristiana Belulescu, Claudiu Mărgăritescu, Cristiana Iulia Dumitrescu, Maria Cristina Munteanu, Luminiţa Dăguci, Otilia Clara Mărgăritescu, Marius Matei
ABSTRACT
Adenoid cystic carcinoma (ACC) is the second most common malignant salivary glands neoplasms with a controversial biological behavior. Even though these tumors grow slowly, they have increased potential for recurrence and distant metastasis. In order to elucidate this behavior, our study aimed to investigate the immunoexpression in such tumors of the most important transcriptional factors [Twist, Snail, Slug, and zinc finger E-box binding homeobox 1 (ZEB1)] involved in the epithelial-mesenchymal transition process. The highest level of expression was recorded for Twist, present in all the investigated cases, followed by the Slug and Snail, while no tumor parenchyma reactivity was noticed for the ZEB1 factor. There were tumor reactivity differences regarding topography, histopathological variant, and nerve and lymph node invasion status. Thus, tumors developed from the intraoral minor salivary glands, with solid pattern, perineural invasion, locally aggressive and with lymph node metastasis were the most reactive. Therefore, these transcription factors could be useful as prognostic biomarkers and efficient therapeutic targets in such salivary malignancies.
Sigma-Aldrich
Monoclonal Anti-ZEB1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0151, purified immunoglobulin, buffered aqueous glycerol solution