MilliporeSigma
  • Circulating fibroblast growth factor-2 precipitates HIV nephropathy in mice.
Circulating fibroblast growth factor-2 precipitates HIV nephropathy in mice.
Disease models & mechanisms (2021-07-27)
Jharna R Das, Marina Jerebtsova, Pingtao Tang, Jinliang Li, Jing Yu, Patricio E Ray
ABSTRACT
People of African ancestry living with the human immunodeficiency virus-1 (HIV-1) are at risk of developing HIV-associated nephropathy (HIVAN). Children with HIVAN frequently show high plasma fibroblast growth factor-2 (FGF-2) levels; however, the role of circulating FGF-2 in the pathogenesis of childhood HIVAN is unclear. Here, we explored how circulating FGF-2 affected the outcome of HIVAN in young HIV-Tg26 mice. Briefly, we demonstrated that FGF-2 was preferentially recruited in the kidneys of mice without pre-existing kidney disease, precipitating HIVAN by activating phosphorylated extracellular signal-regulated kinase (pERK) in renal epithelial cells, without inducing the expression of HIV-1 genes. Wild-type mice injected with recombinant adenoviral FGF-2 (rAd-FGF-2) vectors carrying a secreted form of human FGF-2 developed transient and reversible HIVAN-like lesions, including proteinuria and glomerular enlargement. HIV-Tg26 mice injected with rAd-FGF-2 vectors developed more-significant proliferative and pro-fibrotic inflammatory lesions, similar to those seen in childhood HIVAN. These lesions were partially reversed by treating mice with the FGF/VEGF receptor tyrosine kinase inhibitor PD173074. These findings suggest that high plasma FGF-2 levels may be an independent risk factor for precipitating HIVAN in young children.
Sigma-Aldrich
FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074, The FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074 controls the biological activity of FGF/VEGF Receptor Tyrosine Kinase. Primarily used for Phosphorylation & Dephosphorylation applications.