MilliporeSigma
  • Discriminatory alteration of carbohydrate homeostasis by gold nanoparticles ingestion in Drosophila.
Discriminatory alteration of carbohydrate homeostasis by gold nanoparticles ingestion in Drosophila.
Toxicology and industrial health (2020-11-27)
Akanksha Raj, Prasanna Shah, Akanksha Singh, Namita Agrawal
ABSTRACT
With the extensive usage of gold nanoparticles (AuNPs) in various industrial sectors and biomedical applications, evaluation of their possible effects on human health becomes imperative. Therefore, the present study was aimed toward assessing the dose-dependent impact of AuNPs ingestion on metabolic homeostasis using Drosophila melanogaster as a model system. We found that larval ingestion of higher dose of AuNPs significantly reduced body weight. Further analysis of the crucial energy reservoir showed selective alteration in carbohydrate levels without any change in the lipid and protein levels. Transcriptional downregulation of glycogen synthase further supported impaired glycogen metabolism in flies supplemented with higher dose of AuNPs. Additionally, ingestion of higher dose of AuNPs in larvae results in significantly increased levels of reactive oxygen species (ROS) in the peripheral tissues, suggestive of stress condition. Our findings clearly imply that supplementing higher doses of AuNPs at an early developmental stage can potentially cause weight loss, impair glycogen metabolism, and elevate ROS production. Therefore, determination of a biologically effective dose is critical for the safety of mankind and vulnerable populations at the workplace.
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%