MilliporeSigma
PTEN loss correlates with T cell exclusion across human cancers.
BMC cancer (2021-04-21)
Ziying Lin, Lixia Huang, Shao Li Li, Jincui Gu, Xiaoxian Cui, Yanbin Zhou
ABSTRACT
Recent evidences had shown that loss in phosphatase and tensin homolog deleted on chromosome 10 (PTEN) was associated with immunotherapy resistance, which may be attributed to the non-T-cell-inflamed tumor microenvironment. The impact of PTEN loss on tumor microenvironment, especially regarding T cell infiltration across tumor types is not well understood. Utilizing The Cancer Genome Atlas (TCGA) and publicly available dataset of immunotherapy, we explored the correlation of PTEN expressing level or genomic loss with tumor immune microenvironment and response to immunotherapy. We further investigated the involvement of PI3K-AKT-mTOR pathway activation, which is known to be the subsequent effect of PTEN loss, in the immune microenvironment modulation. We reveal that PTEN mRNA expression is significantly positively correlated with CD4/CD8A gene expression and T cells infiltration especially T helpers cells, central memory T cell and effector memory T cells in multiples tumor types. Genomic loss of PTEN is associated with reduced CD8+ T cells, type 1 T helper cells, and increased type 2 T helper cells, immunosuppressed genes (e.g. VEGFA) expression. Furthermore, T cell exclusive phenotype is also observed in tumor with PI3K pathway activation or genomic gain in PIK3CA or PIK3CB. PTEN loss and PI3K pathway activation correlate with immunosuppressive microenvironment, especially in terms of T cell exclusion. PTEN loss predict poor therapeutic response and worse survival outcome in patients receiving immunotherapy. These data brings insight into the role of PTEN loss in T cell exclusion and immunotherapy resistance, and inspires further research on immune modulating strategy to augment immunotherapy.