MilliporeSigma
  • Determination of chlorzoxazone and 6-hydroxychlorzoxazone in plasma by gas chromatography--mass spectrometry.
Determination of chlorzoxazone and 6-hydroxychlorzoxazone in plasma by gas chromatography--mass spectrometry.
Journal of chromatography. B, Biomedical sciences and applications (1998-03-14)
C B Eap, C Schnyder, L Savary
ABSTRACT
A gas chromatographic-mass spectrometric method is presented which allows the determination of chlorzoxazone and 6-hydroxychlorzoxazone after derivatization with the reagent N-tert.-butyldimethylsilyl-N-methyltrifluoroacetamide. No interference was observed from endogenous compounds following the extraction of plasma samples from six different human subjects. The standard curves were linear over a working range of 20 to 4000 ng/ml and of 20 to 1000 ng/ml for chlorzoxazone and 6-hydroxychlorzoxazone, respectively. Recoveries ranged from 65 to 97% for the two compounds and intra- and inter-day coefficients of variation were always less than 9%. The limit of quantitation of the method was found to be 5 ng/ml for the two compounds, hence allowing its use for single low dose pharmacokinetics.
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)