MilliporeSigma
  • TBX3 acts as tissue-specific component of the Wnt/β-catenin transcriptional complex.
TBX3 acts as tissue-specific component of the Wnt/β-catenin transcriptional complex.
eLife (2020-08-19)
Dario Zimmerli, Costanza Borrelli, Amaia Jauregi-Miguel, Simon Söderholm, Salome Brütsch, Nikolaos Doumpas, Jan Reichmuth, Fabienne Murphy-Seiler, MIchel Aguet, Konrad Basler, Andreas E Moor, Claudio Cantù
ABSTRACT
BCL9 and PYGO are β-catenin cofactors that enhance the transcription of Wnt target genes. They have been proposed as therapeutic targets to diminish Wnt signaling output in intestinal malignancies. Here we find that, in colorectal cancer cells and in developing mouse forelimbs, BCL9 proteins sustain the action of β-catenin in a largely PYGO-independent manner. Our genetic analyses implied that BCL9 necessitates other interaction partners in mediating its transcriptional output. We identified the transcription factor TBX3 as a candidate tissue-specific member of the β-catenin transcriptional complex. In developing forelimbs, both TBX3 and BCL9 occupy a large number of Wnt-responsive regulatory elements, genome-wide. Moreover, mutations in Bcl9 affect the expression of TBX3 targets in vivo, and modulation of TBX3 abundance impacts on Wnt target genes transcription in a β-catenin- and TCF/LEF-dependent manner. Finally, TBX3 overexpression exacerbates the metastatic potential of Wnt-dependent human colorectal cancer cells. Our work implicates TBX3 as context-dependent component of the Wnt/β-catenin-dependent transcriptional complex.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HA Tag Antibody, clone 114-2C-7, rabbit monoclonal, clone 114-2C-7, Upstate®, from rabbit