MilliporeSigma
  • 5,5-Dialkylluciferins are thermal stable substrates for bioluminescence-based detection systems.
5,5-Dialkylluciferins are thermal stable substrates for bioluminescence-based detection systems.
PloS one (2020-12-15)
Ce Shi, Michael P Killoran, Mary P Hall, Paul Otto, Monika G Wood, Ethan Strauss, Lance P Encell, Thomas Machleidt, Keith V Wood, Thomas A Kirkland
ABSTRACT
Firefly luciferase-based ATP detection assays are frequently used as a sensitive, cost-efficient method for monitoring hygiene in many industrial settings. Solutions of detection reagent, containing a mixture of a substrate and luciferase enzyme that produces photons in the presence of ATP, are relatively unstable and maintain only a limited shelf life even under refrigerated conditions. It is therefore common for the individual performing a hygiene test to manually prepare fresh reagent at the time of monitoring. To simplify sample processing, a liquid detection reagent with improved thermal stability is needed. The engineered firefly luciferase, Ultra-Glo™, fulfills one aspect of this need and has been valuable for hygiene monitoring because of its high resistance to chemical and thermal inactivation. However, solutions containing both Ultra-Glo™ luciferase and its substrate luciferin gradually lose the ability to effectively detect ATP over time. We demonstrate here that dehydroluciferin, a prevalent oxidative breakdown product of luciferin, is a potent inhibitor of Ultra-Glo™ luciferase and that its formation in the detection reagent is responsible for the decreased ability to detect ATP. We subsequently found that dialkylation at the 5-position of luciferin (e.g., 5,5-dimethylluciferin) prevents degradation to dehydroluciferin and improves substrate thermostability in solution. However, since 5,5-dialkylluciferins are poorly utilized by Ultra-Glo™ luciferase as substrates, we used structural optimization of the luciferin dialkyl modification and protein engineering of Ultra-Glo™ to develop a luciferase/luciferin pair that shows improved total reagent stability in solution at ambient temperature. The results of our studies outline a novel luciferase/luciferin system that could serve as foundations for the next generation of bioluminescence ATP detection assays with desirable reagent stability.
Sigma-Aldrich
6-Hydroxybenzothiazole-2-carbonitrile, 96%
Sigma-Aldrich
DL-Cysteine, technical grade