MilliporeSigma
  • Propofol Protects Against TNF-α-induced Blood-brain Barrier Disruption via the PIM-1/eNOS/NO Pathway.
Propofol Protects Against TNF-α-induced Blood-brain Barrier Disruption via the PIM-1/eNOS/NO Pathway.
Current neurovascular research (2020-08-21)
Yan Lu, Zhendong Xu, Fuyi Shen, Rong Lin, Haibing Li, Xiang Lv, Zhiqiang Liu
ABSTRACT
The Inflammatory cytokine, tumor necrosis factor-α (TNF-α), disrupts blood-brain barrier (BBB). Propofol reportedly exerts an anti-inflammatory effect in the central nervous system. We hypothesized that propofol could provide a protective effect against TNF-α-induced disruption in human cerebral microvascular endothelial cells (hCMEC/D3 cells) and explored the underlying mechanisms. The hCMEC/D3 cell monolayers were pretreated with propofol, followed by TNF-α treatment. The integrity of BBB was reflected by assessing the trans-endothelial electrical resistance (TEER) and determining the expression of proteins within tight junctions (TJs). The effect of propofol on TNF-α-modulated nitric oxide production was measured by a nitrate reductase assay kit. The expression of ZO-1, claudin-5, occludin, TNF receptor 1 (TNFR1), TNF receptor 2 (TNFR2), proviral-integration site for Moloney murine leukaemia virus (PIM)-1kinase, the phosphorylation of endothelial nitric oxide synthase at ser633 (peNOS-ser633) were detected by western blot. In hCMEC/D3 cells, TNF-α treatment markedly disrupted the integrity of BBB. Further, we found TNF-α treatment could increase the expression of PIM-1, then activate the phosphorylation of eNOS and induce the release of nitric oxide (NO). More importantly, we found that TNF- α-impaired BBB integrity could be reversed by propofol. These results suggest that the PIM-1/eNOS/NO pathway plays a vital role, in which Propofol protects against TNF-α-induced blood-brain barrier disruption.