MilliporeSigma
  • Biodistribution and intracellular localization of hyaluronan and its nanogels. A strategy to target intracellular S. aureus in persistent skin infections.
Biodistribution and intracellular localization of hyaluronan and its nanogels. A strategy to target intracellular S. aureus in persistent skin infections.
Journal of controlled release : official journal of the Controlled Release Society (2020-06-20)
E Montanari, P Mancini, F Galli, M Varani, I Santino, T Coviello, L Mosca, P Matricardi, F Rancan, C Di Meo
ABSTRACT
Intracellular pathogens are a critical challenge for antimicrobial therapies. Staphylococcus aureus (S. aureus) causes approximately 85% of all skin and soft tissue infections in humans worldwide and more than 30% of patients develop chronic or recurrent infections within three months, even after appropriate antibacterial therapies. S. aureus is also one of the most common bacteria found in chronic wounds. Recent evidences suggest that S. aureus is able to persist within phagolysosomes of skin cells (i.e. keratinocytes, phagocytic cells), being protected from both the immune system and a number of antimicrobials. To overcome these limits, nano-formulations that enable targeted therapies against intracellular S. aureus might be developed. Herein, the biodistribution and intracellular localisation of hyaluronan (HA) and HA-based nanoparticles (nanogels, NHs) are investigated, both after intravenous (i.v.) injections (in mice) and topical administrations (in ex vivo human skin). Results indicate HA and NHs accumulate especially in skin and liver of mice after i.v. injection. After topical application on human skin explants, no penetration of both HA and NHs was detected in skin with intact stratum corneum. By contrast, in barrier-disrupted human skin (with partial removal and loosening of stratum corneum), HA and NHs penetrate to the viable epidermis and are taken up by keratinocytes. In mechanically produced wounds (skin without epidermis) they accumulate in wound tissue and are taken up by dermis cells, e.g. fibroblasts and phagocytic cells. Interestingly, in all cases, the cellular uptake is CD44-mediated. In vitro studies confirmed that after CD44-mediated uptake, both HA and NHs accumulate in lysosomes of dermal fibroblasts and macrophages, as previously reported for keratinocytes. Finally, the colocalisation between intracellular S. aureus and HA or NHs is demonstrated, in macrophages. Altogether, for the first time, these results strongly suggest that HA and HA-based NHs can provide a targeted therapy to intracellular S. aureus, in persistent skin or wound infections.
Sigma-Aldrich
4-Bromobutyric acid, 98%