MilliporeSigma
  • WAY-163909, a 5-HT2C agonist, enhances the preclinical potency of current antipsychotics.
WAY-163909, a 5-HT2C agonist, enhances the preclinical potency of current antipsychotics.
Psychopharmacology (2008-12-25)
Steven M Grauer, Radka Graf, Rachel Navarra, Amy Sung, Sheree F Logue, Gary Stack, Christine Huselton, Zhi Liu, Thomas A Comery, Karen L Marquis, Sharon Rosenzweig-Lipson
ABSTRACT
5-HT(2C) agonists, by decreasing mesolimbic dopamine without affecting nigrostriatal dopamine, are predicted to have antipsychotic efficacy with low extrapyramidal side effects (EPS). Combining 5-HT(2C) agonists with low doses of existing antipsychotics could increase treatment efficacy while reducing treatment liabilities such as EPS (typical antipsychotics), and the propensity for weight gain (atypical antipsychotics). The objectives of these studies were to combine WAY-163909, a selective 5-HT(2C) agonist, with either the typical antipsychotic haloperidol, or the atypical antipsychotic clozapine, at doses that were ineffective on their own, with the expectation that a shift in potency in several rodent behavior models predictive of antipsychotic activity would occur. In mice, co-administration of either haloperidol, or clozapine, produced a significant leftward shift in the ability of WAY-163909 to block apomorphine-induced climbing behavior, without any affect on apomorphine-induced stereotypy or an increased propensity for catalepsy. In the rat-conditioned avoidance model, WAY-163909 was combined with either haloperidol or clozapine at doses that individually produced reductions in avoidance response on the order of 10%, while the combination of WAY-163909 and either of the antipsychotics resulted in a greater than 70% reduction in avoidance, with no evidence of response failures, or pharmacokinetic interaction. Doses of either haloperidol or clozapine, that failed to antagonize an MK-801 induced deficit in prepulse inhibition, significantly attenuated the sensory gating deficit when combined with WAY-163909. Data support the notion that 5-HT(2C) receptor agonists, co-administered with other marketed antipsychotics, allow for dose sparing with a more favorable side-effect profile.
Sigma-Aldrich
WAY-163909 hydrochloride, ≥98% (HPLC)