MilliporeSigma
  • miR-34a-5p Inhibits Cell Proliferation, Migration and Invasion Through Targeting JAG1/Notch1 Pathway in HPV-Infected Human Epidermal Keratinocytes.
miR-34a-5p Inhibits Cell Proliferation, Migration and Invasion Through Targeting JAG1/Notch1 Pathway in HPV-Infected Human Epidermal Keratinocytes.
Pathology oncology research : POR (2019-11-30)
Ying Gao, Ming Yang, Liuliu Wei, Xiaofang Liang, Fang Wu, Yalan Huang, Tao Yang
ABSTRACT
Condyloma acuminate (CA) is a communicable disease caused by human papillomavirus (HPV). This study aimed to study the targeting relationship between miR-34a-5p and Jagged 1 (JAG1), as well as its regulatory effect in HPV-infected cells. Human keratinocyte HaCaT cells were infected with HPV16E6, and CA tissues were collected. The expression level of miR-34a-5p and JAG1 were detected in CA tissues and HPV-HaCaT cells. Cell proliferation, migration and invasion were respectively measured using 3-(4, 5)-dimethylthiahiazo-(-z-y1)-3, 5-diphenytetrazoliumromide (MTT), cell wound healing and Transwell assay. The potential binding sites of miR-34a-5p and JAG1 were predicted by website TargetScan, and confirmed using dual luciferase reporter gene assay. The proteins of Notch1 pathway-related were assessed using western blotting. The results showed that miR-34a-5p expression was decreased, and JAG1 expression was increased in CA tissues and HPV-HaCaT cells. Cell proliferation, migration and invasion were decreased when miR-34a-5p over-expression and JAG1 knock-down in HPV-HaCaT cells. Furthermore, miR-34a-5p had a targeting effect on JAG1. The expression level of Notch1, NICD, Hes1 and Hey1 were increased when miR-34a-5p knock-down. miR-34a-5p could inhibit cell development, and regulate the activity of Notch1 pathway through targeting JAG1 expression in HPV-infected keratinocytes.
Sigma-Aldrich
MISSION® esiRNA, targeting human JAG1