MilliporeSigma
  • NCOA5 is a master regulator of amino acid-induced mTOR activation and β-casein synthesis in bovine mammary epithelial cells.
NCOA5 is a master regulator of amino acid-induced mTOR activation and β-casein synthesis in bovine mammary epithelial cells.
Biochemical and biophysical research communications (2020-08-02)
Xiaohan Yuan, Li Zhang, Yingjun Cui, Yanbo Yu, Xuejun Gao, Jinxia Ao
ABSTRACT
The nuclear receptor co-activator 5 (NCOA5) is known as a co-activator or co-repressor that influences gene expression and cellular physiology, but its roles and detailed molecular mechanism is still largely unknown. In this study, we explored the role and molecular mechanism of NCOA5 in amino acid-induced activation of the mechanistic target of rapamycin (mTOR) and milk protein synthesis in bovine mammary epithelial cells (BMECs). Methionine (Met) and leucine (Leu) significantly up-regulated the expression of NCOA5. NCOA5 overexpression increased mTOR phosphorylation and β-casein synthesis, whereas its knockdown exhibited the opposite effects. Furthermore, inhibition of phosphatidylinositol 3-kinase (PI3K) completely abolished the stimulatory effects of Met and Leu on NCOA5 expression. ChIP-qPCR analysis detected that NCOA5 bound to the mTOR promoter, and this interaction was enhanced by the stimulation of Met and Leu. These above data reveal that NCOA5 is a key regulator of amino acid-induced PI3K-mediated mTOR activation and β-casein synthesis in BMECs.
Sigma-Aldrich
MISSION® esiRNA, targeting human NCOA5