MilliporeSigma
  • Large-Scale Evaluation of Major Soluble Macromolecular Components of Fish Muscle from a Conventional 1H-NMR Spectral Database.
Large-Scale Evaluation of Major Soluble Macromolecular Components of Fish Muscle from a Conventional 1H-NMR Spectral Database.
Molecules (Basel, Switzerland) (2020-04-29)
Feifei Wei, Minoru Fukuchi, Kengo Ito, Kenji Sakata, Taiga Asakura, Yasuhiro Date, Jun Kikuchi
ABSTRACT
Conventional proton nuclear magnetic resonance (1H-NMR) has been widely used for identification and quantification of small molecular components in food. However, identification of major soluble macromolecular components from conventional 1H-NMR spectra is difficult. This is because the baseline appearance is masked by the dense and high-intensity signals from small molecular components present in the sample mixtures. In this study, we introduced an integrated analytical strategy based on the combination of additional measurement using a diffusion filter, covariation peak separation, and matrix decomposition in a small-scale training dataset. This strategy is aimed to extract signal profiles of soluble macromolecular components from conventional 1H-NMR spectral data in a large-scale dataset without the requirement of re-measurement. We applied this method to the conventional 1H-NMR spectra of water-soluble fish muscle extracts and investigated the distribution characteristics of fish diversity and muscle soluble macromolecular components, such as lipids and collagens. We identified a cluster of fish species with low content of lipids and high content of collagens in muscle, which showed great potential for the development of functional foods. Because this mechanical data processing method requires additional measurement of only a small-scale training dataset without special sample pretreatment, it should be immediately applicable to extract macromolecular signals from accumulated conventional 1H-NMR databases of other complex gelatinous mixtures in foods.
Sigma-Aldrich
3-(Trimethylsilyl)-1-propanesulfonic acid-d6 sodium salt, 98 atom % D