MilliporeSigma
  • A biosensor for cholesterol based on gold nanoparticles-catalyzed luminol electrogenerated chemiluminescence.
A biosensor for cholesterol based on gold nanoparticles-catalyzed luminol electrogenerated chemiluminescence.
Biosensors & bioelectronics (2011-12-31)
Meihe Zhang, Ruo Yuan, Yaqin Chai, Shihong Chen, Huaan Zhong, Cun Wang, Yinfeng Cheng
ABSTRACT
A novel cholesterol biosensor was prepared based on gold nanoparticles-catalyzed luminol electrogenerated chemiluminescence (ECL). Firstly, l-cysteine-reduced graphene oxide composites were modified on the surface of a glassy carbon electrode. Then, gold nanoparticles (AuNPs) were self-assembled on it. Subsequently, cholesterol oxidase (ChOx) was adsorbed on the surface of AuNPs to construct a cholesterol biosensor. The stepwise fabrication processes were characterized with cyclic voltammetry and atomic force microscopy. The ECL behaviors of the biosensor were also investigated. It was found that AuNPs not only provided larger surface area for higher ChOx loading but also formed the nano-structured interface on the electrode surface to improve the analytical performance of the ECL biosensor for cholesterol. Besides, based on the efficient catalytic ability of AuNPs to luminol ECL, the response of the biosensor to cholesterol was linear range from 3.3 μM to 1.0 mM with a detection limit of 1.1 μM (S/N=3). In addition, the prepared ECL biosensor exhibited satisfying reproducibility, stability and selectivity. Taking into account the advantages of ECL, we confidently expect that ECL would have potential applications in biotechnology and clinical diagnosis.
Sigma-Aldrich
Cholesterol Oxidase from microorganisms, lyophilized powder, ≥50 units/mg protein, recombinant, expressed in E. coli