MilliporeSigma
  • Facile synthesis of hydrophobic fluoroalkyl functionalized silsesquioxane nanostructures.
Facile synthesis of hydrophobic fluoroalkyl functionalized silsesquioxane nanostructures.
Chemical communications (Cambridge, England) (2007-12-01)
Scott T Iacono, Ashwani Vij, Wade Grabow, Dennis W Smith, Joseph M Mabry
ABSTRACT
New fluorinated polyhedral oligomeric silsesquioxane (F-POSS) structures possessing a high degree of hydrophobicity have been prepared via a facile corner-capping methodology.
Sigma-Aldrich
Trichloro(3,3,3-trifluoropropyl)silane, 97%