MilliporeSigma
  • Characterization of AN6001, a positive allosteric modulator of α6β2-containing nicotinic acetylcholine receptors.
Characterization of AN6001, a positive allosteric modulator of α6β2-containing nicotinic acetylcholine receptors.
Biochemical pharmacology (2019-12-31)
Marloes van Hout, Jessica Klein, Philip K Ahring, David T Brown, Siganya Thaneshwaran, Altair B Dos Santos, Anders A Jensen, Kristi A Kohlmeier, Palle Christophersen, Tino Dyhring
ABSTRACT
α6β2-Containing nicotinic acetylcholine receptors (α6β2* nAChRs) are predominantly expressed in midbrain dopaminergic neurons, including substantia nigra pars compacta (SNc) neurons and their projections to striatal regions, where they regulate dopamine release and nigrostriatal activity. It is well established that nAChR agonists exert protection against dopaminergic neurotoxicity in cellular assays and parkinsonian animal models. Historically, drug development in the nAChR field has been mostly focused on development of selective agonists and positive allosteric modulators (PAMs) for the predominant neuronal nAChRs, α7 and α4β2. Here, we report the discovery and characterization of AN6001, a novel selective α6β2* nAChR PAM. AN6001 mediated increases in both nicotine potency and efficacy at the human α6/α3β2β3V9'S nAChR in HEK293 cells, and it positively modulated ACh-evoked currents through both α6/α3β2β3V9'S and a concatenated β3-α6-β2-α6-β2 receptor in Xenopus oocytes, displaying EC50 values of 0.58 µM and 0.40 µM, respectively. In contrast, the compound did not display significant modulatory activity at α4β2, α3β4, α7 and muscle nAChRs. AN6001 also increased agonist-induced dopamine release from striatal synaptosomes and augmented agonist-induced global cellular responses and inward currents in dopaminergic neurons in SNc slices (measured by Ca2+ imaging and patch clamp recordings, respectively). Finally, AN6001 potentiated the neuroprotective effect of nicotine at MPP+-treated primary dopaminergic neurons. Overall, our studies demonstrate the existence of allosteric sites on α6β2* nAChRs and that positive modulation of native α6β2* receptors strengthens DA signaling. Hence, AN6001 represents an important tool for studies of α6β2* nAChRs and furthermore underlines the therapeutic potential in these receptors in Parkinson's disease.