MilliporeSigma
  • Targeted photoimmunotherapy based on photosensitizer-antibody conjugates for multiple myeloma treatment.
Targeted photoimmunotherapy based on photosensitizer-antibody conjugates for multiple myeloma treatment.
Journal of photochemistry and photobiology. B, Biology (2020-01-14)
Wael M Darwish, Noha A Bayoumi, Hanan M El-Shershaby, Nasr M Allahloubi
ABSTRACT
Despite the high in vitro efficacy of photodynamic therapeutics, lack of tumor targeting significantly reduces their in vivo efficacy and thus limits their clinical use. Photoimmunotherapy (PIT) is a new synthetic strategy to target and treat cancer by photodynamic therapy (PDT). In this study, we describe design and synthesis of a third-generation photosensitizer comprising a PEGylated-phthalocyanine star-polymer photosensitizer that covalently bound to a myeloma tumor-selective antibody (MAb) via the carbodiimide chemistry. The free photosensitizer demonstrated a minimum dark toxicity when tested in mammalian myeloma cell line (SP2/OR); and a moderate phototoxicity after irradiation with non thermal laser red light as a result of light-induced production of cytotoxic singlet oxygen species. Covalent attachment of the photosensitizer (Pc) to the MAb resulted in a significantly enhanced phototoxicity. This is mainly ascribed to the fact that internalization enhances phototoxicity of Pc-MAb bioconjugates. The radioactivated photoimmuno-conjugates 131I(PcMAb) demonstrated the highest phototoxicity to myeloma cells. The suggested bioconjugates are promising candidates as multiple therapeutic models for in vivo treatment of myeloma.
Sigma-Aldrich
4-Nitrophthalonitrile, 99%