MilliporeSigma
  • Downregulation of the polyamine regulator spermidine/spermine N(1)-acetyltransferase by Epstein-Barr virus in a Burkitt's lymphoma cell line.
Downregulation of the polyamine regulator spermidine/spermine N(1)-acetyltransferase by Epstein-Barr virus in a Burkitt's lymphoma cell line.
Virus research (2013-07-31)
Mingxia Shi, Yan-Jun Gan, Timothy O Davis, Rona S Scott
ABSTRACT
Transition of Akata Burkitt's lymphoma (BL) from a malignant to nonmalignant phenotype upon loss of Epstein-Barr virus (EBV) is evidence for a viral contribution to tumorigenesis despite the tight restriction of EBV gene expression in BL. Examination of global cellular gene expression in Akata subclones that retained or lost EBV identified spermidine/spermine N(1)-acetyltransferase (SAT1), an inducible enzyme whose catabolism of polyamines affects both apoptosis and cell growth, as one of a limited number of cellular genes downregulated by EBV. Re-infection of the EBV-negative Akata clone reduced SAT1 mRNA to a level comparable with the parental EBV-positive Akata. EBV-positive Akata cells demonstrated decreased SAT1 enzyme activity concomitant with altered intracellular polyamine constituents. Reduction of SAT1 in EBV-positive BL was a transcriptional effect. Forced expression of the viral BCL2 homologue, BHRF1, in an EBV-negative Akata clone reduced SAT1 mRNA. Thus, EBV repression of polyamine catabolism becomes a complementary alteration to dysregulated c-myc enhancement of polyamine synthesis in BL and favorable to BL lymphomagenesis.
Sigma-Aldrich
Anti-EBV EA-R-p17 Antibody, clone 5B11, clone 5B11, Chemicon®, from mouse