MilliporeSigma
  • Naloxone blockade of myocardial ischemic preconditioning does not require central nervous system participation.
Naloxone blockade of myocardial ischemic preconditioning does not require central nervous system participation.
Basic research in cardiology (1999-05-18)
G L Chien, K Mohtadi, R A Wolff, D M Van Winkle
ABSTRACT
The hypothesis that naloxone blockade of ischemic preconditioning (IP)-induced infarct limitation does not require central nervous system participation was evaluated using quaternary naloxone in anesthetized rabbits (Study I) and naloxone hydrochloride in isolated rabbit hearts (Study II). In Study I, rabbits underwent 30 min coronary artery occlusion and 180 min reperfusion. IP was elicited with a 5 min coronary artery occlusion beginning 15 min before the 30 min occlusion. Intravenous naloxone methiodide, 12.9 mg/kg, was bolused 10 or 1 min before IP. In Study II, rabbit hearts underwent 45 min coronary artery occlusion and 120 min reperfusion. IP was elicited with 2 cycles of 5 min coronary artery occlusion plus 5 min reperfusion, beginning 20 min before the 45 min occlusion. Naloxone hydrochloride, 1 mumol/L or 100 mumol/L, was added to the buffer perfusate for 25 min preceding the long coronary artery occlusion. In both studies, infarct size was assessed with tetrazolium, normalized to risk volume, and analyzed using ANOVA. In both studies, IP reduced infarct size compared to control (6.3 +/- 2.3 vs. 29.5 +/- 4.4, P = 0.007, Study I; 11.8 +/- 4.7 vs. 47.7 +/- 6.7, P = 0.03, Study II). In Study I, IP was not blocked when naloxone methiodide was given 10 min before IP (13.8 +/- 4.8 vs. 42.3 +/- 5.4, P = 0.004) but was blocked when given 1 min before IP (25.3 +/- 7.2 vs. 28.4 +/- 5.0, P = ns). In Study II, infarct size was intermediate in the 1 mumol/L naloxone hydrochloride + IP group (19.0 +/- 6.5 vs. 48.9 +/- 8.4, P = ns) but IP was blocked by 100 mumol/L naloxone hydrochloride (62.6 +/- 4.5 vs. 56.2 +/- 6.7, P = ns). Naloxone blockade of IP-induced infarct limitation involves a cardiac mechanism.
Sigma-Aldrich
Naloxone methiodide, ≥98% (HPLC), solid