MilliporeSigma
  • Catalytic Carbonylative Rearrangement of Norbornadiene via Dinuclear Carbon-Carbon Oxidative Addition.
Catalytic Carbonylative Rearrangement of Norbornadiene via Dinuclear Carbon-Carbon Oxidative Addition.
Journal of the American Chemical Society (2017-09-19)
Douglas R Hartline, Matthias Zeller, Christopher Uyeda
ABSTRACT
Single bonds between carbon atoms are inherently challenging to activate using transition metals; however, ring-strain release can provide the necessary thermodynamic driving force to make such processes favorable. In this report, we describe a strain-induced C-C oxidative addition of norbornadiene. The reaction is mediated by a dinuclear Ni complex, which also serves as a catalyst for the carbonylative rearrangement of norbornadiene to form a bicyclo[3.3.0] product.
Sigma-Aldrich
iPr-NDI, ≥95%