MilliporeSigma
  • Competitive inhibition by adenosine 5'-triphosphate of the actions on human platelets of 2-chloroadenosine 5'-diphosphate, 2-azidoadenosine 5'-diphosphate and 2-methylthioadenosine 5'-diphosphate.
Competitive inhibition by adenosine 5'-triphosphate of the actions on human platelets of 2-chloroadenosine 5'-diphosphate, 2-azidoadenosine 5'-diphosphate and 2-methylthioadenosine 5'-diphosphate.
British journal of pharmacology (1982-10-01)
N J Cusack, S M Hourani
ABSTRACT
1 Adenosine 5'-diphosphate (ADP) induces human platelet aggregation and noncompetitively inhibits stimulated human platelet adenylate cyclase; these two effects are mediated by the same ADP receptor, at which adenosine 5'-triphosphate (ATP) is a competitive antagonist. 2 Two ADP analogues, 2-azidoadenosine 5'-diphosphate (2-azido-ADP) and 2-methylthioadenosine 5'-diphosphate (2-methylthio-ADP) have been reported to be more potent as inhibitors of adenylate cyclase than they are as aggregating agents, but no evidence has been presented that these actions are mediated solely by the ADP receptor. 3 We therefore tested the ability of ATP to inhibit the actions of these compounds and of another ADP analogue, 2-chloroadenosine 5'-diphosphate (2-chloro-ADP). 4 2-Chloro-ADP, 2-azido-ADP and 2-methylthio-ADP each induced aggregation and inhibited stimulated adenylate cyclase. Both of these actions were competitively inhibited by ATP (50 microM) with pA2 values similar to those previously found for inhibition by ATP of these effects of ADP. 5 The reported greater potency of 2-azido-ADP and of 2-methylthio-ADP as inhibitors of adenylate cyclase than as aggregating agents is therefore due only to their greater efficacy for this effect, not to some extra actions elsewhere.