MilliporeSigma
  • A non-isotopic assay uses bromouridine and RNA synthesis to detect DNA damage responses.
A non-isotopic assay uses bromouridine and RNA synthesis to detect DNA damage responses.
Mutation research (2010-04-17)
Mayu Hasegawa, Shigenori Iwai, Isao Kuraoka
ABSTRACT
Individuals with inherited xeroderma pigmentosum (XP) disorder and Cockayne syndrome (CS) are deficient in nucleotide excision repair and experience hypersensitivity to sunlight. Although there are several diagnostic assays for these disorders, the recovery of RNA synthesis (RRS) assay that can discriminate between XP cells and CS cells is very laborious. Here, we report on a novel non-radioisotope RRS assay that uses bromouridine (a uridine analog) as an alternative to (3)H-uridine. This assay can easily detect RNA polymerase I transcription in nucleoli and RNA polymerase II transcription in nuclei. The non-RI RSS assay also can rapidly detect normal RRS activity in HeLa cells. Thus, this assay is useful as a novel and easy technique for CS diagnosis. Because RRS is thought to be related to transcription-coupled DNA repair, which is triggered by the blockage of transcriptional machinery by DNA lesions, this assay may be of use for analysis of DNA repair, transcription, and/or genetic toxicity.
Sigma-Aldrich
5-Bromouridine, 98%