MilliporeSigma
  • Inhibition of p38 kinase mimics survival signal-linked protection against apoptosis in rat cerebellar granule neurons.
Inhibition of p38 kinase mimics survival signal-linked protection against apoptosis in rat cerebellar granule neurons.
Cellular & molecular biology letters (2001-09-07)
R Nath, K McGinnis, S Dutta, B Shivers, K K Wang
ABSTRACT
The mitogen-activated protein kinase (MAPK) cascades are thought to be important mediators in the transduction of extracellular signals into cellular responses. The p38 kinase, a member of the MAPK superfamily, is activated by a wide variety of extracellular stimuli and has been implicated in neuronal apoptosis induced by glutamate. In this study we have examined the role of p38 kinase in the potassium deprivation model of apoptosis in rat cerebellar granule neurons (CGN). An increase in p38 kinase activity was observed with a 15-minute potassium deprivation when compared to the basal level. We also found that SB203580 and PD169316, specific p38 kinase inhibitors, significantly attenuated apoptosis in potassium-deprived cells in a dose dependent manner. A decrease in caspase-3 mediated DEVD-MCA, substrate hydrolysis and the appearance of the 120 kDa-spectrin breakdown product in cells treated with SB203580 further suggests that the p38 kinase acts upstream of caspase-3 in the apoptosis cascade. The data provides evidence for an essential role of p38 kinase in mediating apoptotic cell death in CGN and the inhibition of p38 kinase mimics the suppression of apoptosis provided by natural survival signals.