MilliporeSigma
  • Development and preliminary validation of a mandarin Chinese language questionnaire measuring betel quid dependency among adults in Taiwan.
Development and preliminary validation of a mandarin Chinese language questionnaire measuring betel quid dependency among adults in Taiwan.
Psychiatry research (2018-11-27)
Ming-Hsui Tsai, Tsai-Chung Li, Chia-Ing Li, Ellen R Gritz, Cho Lam, Chang-Fang Chiu, Chiu-Shong Liu, Chien-Chen Lai, Wei-Yi Hsu, Irene Tamí-Maury, I-Ying Lu, Cheng-Chieh Lin
ABSTRACT
The purposes of this study were to develop the Chinese-version betel quid dependence instrument (BQDI) and to test its reliability and validity. An item pool relevant to betel quid dependence was generated. A panel of three experts assessed content validity including content relevance, clarity, and domain coverage. A cross-sectional study was conducted, consisting of 113 participants from a construction site, betel quid stalls, and a teaching hospital in Taichung, Taiwan. Construct validity was assessed by hypothesizing a significant correlation between the BQDI score and number of pieces-years for betel quid chewing and betel quid biomarkers. The overall Cronbach's alpha coefficient was 0.94. Factor analysis indicated the BQDI consisted of a three-factor structure, including physical and psychological cravings, lack of resistance to betel quid, and maladaptive use. We observed significant associations of BQDI total and factor scores with arecaidine (adjusted odds ratio [OR] for medium total BQDI score: 12.87, 95% CI: 1.45-114.5; high total BQDI score: 28.9, 3.53-236.6) and N-methylnipecotate (medium total BQDI score: 6.18, 1.21-31.62; high total BQDI score: 13.10, 2.72-63.03, respectively). Our results provide preliminary good internal consistency and construct validation of the Chinese-version BQDI as a measure of betel quid dependence in community adults.