MilliporeSigma
  • Cyclic(Alkyl)(Amino)Carbene (CAAC)-Supported Zn Alkyls: Synthesis, Structure and Reactivity in Hydrosilylation Catalysis.
Cyclic(Alkyl)(Amino)Carbene (CAAC)-Supported Zn Alkyls: Synthesis, Structure and Reactivity in Hydrosilylation Catalysis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-03-26)
Jean-Charles Bruyere, David Specklin, Christophe Gourlaouen, Rosita Lapenta, Luis F Veiros, Alfonso Grassi, Stefano Milione, Laurent Ruhlmann, Corinne Boudon, Samuel Dagorne
ABSTRACT
The reactivity of ZnII dialkyl species ZnMe2 with a cyclic(alkyl)(amino)carbene, 1-[2,6-bis(1-methylethyl)phenyl]-3,3,5,5-tetramethyl-2-pyrrolidinylidene (CAAC, 1), was studied and extended to the preparation of robust CAAC-supported ZnII Lewis acidic organocations. CAAC adduct of ZnMe2 (2), formed from a 1:1 mixture of 1 and ZnMe2 , is unstable at room temperature and readily undergoes a CAAC carbene insertion into the Zn-Me bond to produce the ZnX2 -type species (CAAC-Me)ZnMe (3), a reactivity further supported by DFT calculations. Despite its limited stability, adduct 2 was cleanly ionized to robust two-coordinate (CAAC)ZnMe+ cation (5+ ) and derived into (CAAC)ZnC6 F5+ (7+ ), both isolated as B(C6 F5 )4- salts, showing the ability of CAAC for the stabilization of reactive [ZnMe]+ and [ZnC6 F5 ]+ moieties. Due to the lability of the CAAC-ZnMe2 bond, the formation of bis(CAAC) adduct (CAAC)2 ZnMe+ cation (6+ ) was also observed and the corresponding salt [6][B(C6 F5 )4 ] was structurally characterized. As estimated from experimental and calculations data, cations 5+ and 7+ are highly Lewis acidic species and the stronger Lewis acid 7+ effectively mediates alkene, alkyne and CO2 hydrosilylation catalysis. All supporting data hints at Lewis acid type activation-functionalization processes. Despite a lower energy LUMO in 5+ and 7+ , their observed reactivity is comparable to those of N-heterocyclic carbene (NHC) analogues, in line with charge-controlled reactions for carbene-stabilized ZnII organocations.