MilliporeSigma
  • Abdominal Ultrasound for the Diagnosis of Tuberculosis Among Human Immunodeficiency Virus-Positive Inpatients With World Health Organization Danger Signs.
Abdominal Ultrasound for the Diagnosis of Tuberculosis Among Human Immunodeficiency Virus-Positive Inpatients With World Health Organization Danger Signs.
Open forum infectious diseases (2019-04-24)
Rulan Griesel, Karen Cohen, Marc Mendelson, Gary Maartens
ABSTRACT
Studies of the value of abdominal ultrasound for diagnosing human immunodeficiency virus (HIV)-associated tuberculosis have major limitations. We conducted a prospective study of HIV-positive inpatients with cough and World Health Organization danger signs. The reference standard was positive Mycobacterium tuberculosis culture from any site. Participants had at least 2 sputa and 1 blood specimen sent for mycobacterial cultures. Standardized data capture sheets were used for ultrasound reports. A blinded radiologist interpreted chest radiographs, categorized as "likely", "possible", and "unlikely" for HIV-associated tuberculosis. We enrolled 377 participants: 249 women, median age 35 years, 201 with tuberculosis, and median CD4 count 75 cells/µL. The following abdominal ultrasound findings independently predicted tuberculosis: lymph node long-axis ≥10 mm (adjusted odds ratio [aOR], 4.76; 95% confidence interval [CI], 2.41-9.38), splenic hypoechoic lesions (aOR, 3.45; 95% CI, 1.91-6.24), and abdominal/pleural/pericardial effusions (aOR, 1.95; 95% CI, 1.16-3.29). Presence of ≥1 of these 3 features had a sensitivity of 76.4% (95% CI, 69.8-82.3), a specificity of 68.6% (95% CI, 61.1-75.4), and a c-statistic of 0.784 (95% CI, 0.739-0.830). The sensitivity and specificity of chest radiograph assessed as likely tuberculosis was 55.2% (95% CI, 47.2-62.9) and 83.9% (95% CI, 77.0-89.4), respectively. Three features of tuberculosis on abdominal ultrasound independently predicted tuberculosis with moderate diagnostic performance in seriously ill HIV-positive inpatients. Abdominal ultrasound was more sensitive but less specific than chest radiograph for diagnosing tuberculosis in this patient population.