MilliporeSigma
  • In vitro and in silico hormonal activity studies of di-(2-ethylhexyl)terephthalate, a di-(2-ethylhexyl)phthalate substitute used in medical devices, and its metabolites.
In vitro and in silico hormonal activity studies of di-(2-ethylhexyl)terephthalate, a di-(2-ethylhexyl)phthalate substitute used in medical devices, and its metabolites.
Journal of applied toxicology : JAT (2019-03-09)
Nicolas K Kambia, Isabelle Séverin, Amaury Farce, Emmanuel Moreau, Laurence Dahbi, Claire Duval, Thierry Dine, Valérie Sautou, Marie-Christine Chagnon
ABSTRACT
Plasticizers added to polyvinylchloride used in medical devices can be released into patients' biological fluids. The substitution of di-(2-ethylhexyl)phthalate (DEHP) by alternative plasticizers is essential but their safety must be demonstrated. DEHP, di-(2-ethylhexyl)terephthalate (DEHT) and their metabolites were investigated using level 2 Organization for Economic Co-operation and Development bioassays to screen for in vitro hormonal changes. Differences between the DEHP and DEHT metabolites were observed. Albeit weak, the hormonal activities of DEHT-derived metabolites, e.g., 5-OH metabolite of mono-(ethylhexyl)terephthalate (5-OH-MEHT), were detected and the results of docking experiments performed on estrogen receptor alpha and androgen receptor agreed with the biological results. A co-stimulation of human estrogen receptor alpha and human androgen receptor was also observed. With regard to steroidogenesis, a 16-fold increase in estrogen synthesis was measured with 5-OH-MEHT. Therefore, even if DEHT remains an interesting alternative to DEHP because of its low migration from medical devices, it seems important to verify that multi-exposed patients in neonatal intensive care units do not have urinary levels of oxidized metabolites, in particular 5-OH-MEHT, suggesting a potential endocrine-disrupting effect.