MilliporeSigma
  • Identification of Anti-Mycobacterial Biofilm Agents Based on the 2-Aminoimidazole Scaffold.
Identification of Anti-Mycobacterial Biofilm Agents Based on the 2-Aminoimidazole Scaffold.
ChemMedChem (2019-03-06)
T Vu Nguyen, Bradley M Minrovic, Roberta J Melander, Christian Melander
ABSTRACT
Tuberculosis (TB) remains a significant global health problem for which new therapeutic options are sorely needed. The ability of the causative agent, Mycobacterium tuberculosis, to reside within host macrophages and form biofilm-like communities contributes to the persistent and drug-tolerant nature of the disease. Compounds that can prevent or reverse the biofilm-like phenotype have the potential to serve alongside TB antibiotics to overcome this tolerance, and decrease treatment duration. Using Mycobacterium smegmatis as a surrogate organism, we report the identification of two new 2-aminoimidazole compounds that inhibit and disperse mycobacterial biofilms, work synergistically with isoniazid and rifampicin to eradicate preformed M. smegmatis biofilms in vitro, are nontoxic toward Galleria mellonella, and exhibit stability in mouse plasma.